Learn More
Gosselin™ Screw Cap Dippers with Removable Handle
GREENER_CHOICE
Dip with confidence. Gosselin™ Screw Cap Dippers with Removable Handle prevent spillage with screw caps and detachable shaft.
Brand:  Gosselin™ TAT30C-01
403.99 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 15488794
  • £809.00 / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant
  • Packaged individually in tamper evident single zip bags
  • Traceability information on each bag
  • Sterile SAL 10-3
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Sterile SAL 10-3
43 mm
290 mm including container
40 mL
Blue
80 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gosselin™ Screw Cap Dippers with Removable Handle >