Learn More
Eppendorf™ Sealing Mat for DWP96/2000 Deepwell Plate
GREENER_CHOICE
High-performance sealing option provides a secure and reliable closure suitable for incubation, processing, storage, or sample archiving.
Brand:  Eppendorf™ 0030127960
153.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15491920
  • £161.64 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Storage film and foil-glued seals for simple and dependable sealing during sample storage
  • Heat sealing rilm and foil for continuous locking with the best evaporation protection
  • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf plate lid: stable, flexible protection of samples during short-term storage
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCR clean
500/Pk., 5 Bags x 10 Pieces
96/2000 Deep Well Plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Sealing Mat for DWP96/2000 Deepwell Plate > For use with DWP 96/2000 Deep Well Plate