missing translation for 'onlineSavingsMsg'
Learn More

Steris™ Self-Sealing sterilization Pouches

Provides an effective means of sterilization, safe handling and storage of all items until the moment they are used. Steris Corporation™ Self-Sealing sterilization Pouches are CE marked for steam and EO gas.

Brand:  Steris™ NWKP0927

 View more versions of this product

Product Code. 15428782

  • £43.85 / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Self-sealing bags eliminate the need for sealing devices
  • Excellent adhesive strength resistant to steam exposure
  • Non-toxic water-based indicator inks
  • Perforated flap for easy closing
Specifications

Specifications

CE Marked
200/Pk.
90 mm
Steam and Gas EO Sterilizers
90 x 270mm
270 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Steris™ Self-Sealing sterilization Pouches > 90 x 270mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.