missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Signal Boxes

Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.

Brand:  SymLine™ 108160

 View more versions of this product

Product Code. 17863520

This item is not returnable. View return policy

Description

Description

  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad
Specifications

Specifications

Signalbox T1
7.1 in.
HDPE-EC
180 x 105 x 55 mm
Monitoring Filling Levels
SCAT™ Sensors and Safety Waste Caps
3 m
UK-style
220/230 V
Disc Sensor Set, T1, Alarm at Full State, 3 m Signal Cable, 2 m Hook and Loop Fastener
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SymLine™ Signal Boxes > Signal Boxes; Includes: Disc Sensor Set, T1, Alarm at Full State, 3 m Signal Cable, 2 m Hook and Loop Fastener; Length (Metric): 3 m; Type: Signalbox T1; Plug Type: UK-style; Voltage: 220/230 V

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.