missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate 7.5% solution
GREENER_CHOICE

Sodium Bicarbonate 7.5% solution

Brand:  Gibco 25080060

6.76 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10308462

  • £8.10 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Sodium Bicarbonate is buffer commonly used for maintaining the pH of cell culture medium in the presence of 4–10% carbon dioxide. In addition to buffering, sodium bicarbonate provides some nutritional benefit, while rarely displaying any cell toxicity. Formulated as 75 g per liter of water, Gibco™ Sodium Bicarbonate, 7.5% Solution is perfect for supplementing dry powder medium during reconstitution. The final Sodium Bicarbonate concentration used in the medium depends on the media formulation and the carbon dioxide concentration used in the incubator.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Bicarbonate Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Sodium Bicarbonate Solution product made in our Grand Island facility (25080-094). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 to 30°C
7.5 %
Sodium Bicarbonate
100 mL
Sodium Bicarbonate 7.5% Solution, 100mL, 7.5% Concentration, Liquid, Buffer, Used in Mammalian Cell Culture, Perfect for Supplementing Dry Powder Medium During Reconstitution, For Research Use Only, Not for use in Diagnostic Procedures
Liquid
Mammalian Cell Culture
For research use only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Bicarbonate 7.5% solution > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.