Learn More
Cytiva SOURCE™ Tricorn™ Columns for IEX
Source 15Q 4.6/100 PE is a strong anion exchanger prepacked in a Tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
Brand: Cytiva 17-5182-01
Includes: Tricorn columns have Valco fittings for AKTAdesign™ systems and are supplied with M6 connectors for FPLC Systems, instruction book, and two stop plugs attached to the column.
Description
Pre-packed ion exchange columns for high performance ion exchange chromatography of proteins, peptides, polynucleotides and other biomolecules
- 4.6/100 PEEK columns provide fast, high-resolution purifications at lab scale and easy scale-up
- Available with quaternary ammonium strong anion (15Q) or methyl sulfonate strong cation (15S) exhanger
- Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics
- Sample loading up to 25mg of protein/mL
- High stability in organic solvents and at pH extremes
Recommended Storage
4 to 30°C, 20% Ethanol

Specifications
All common ion exchange Buffers | |
1.7mL | |
4.6mm | |
3.93 in. | |
<5 mL/min. | |
Sulphonate group | |
2 to 13 | |
Polystyrene/Divinylbenzene | |
Ion Exchange Column |
Source 15S prepacked column | |
0.18 in. | |
1 x 1.7mL | |
100mm | |
PEEK | |
40 bar | |
15μm | |
4° to 30°C | |
High performance ion exchange chromato graphy of Proteins, Peptides, Polynucleotides and other biomolecules |
Safety and Handling
- Source 15S 4.6/100 PE
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.