Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
For retrieving small, specific volume of powdered or liquid samples
Brand:  Bel-Art™ F36740-0310
 View more versions of this product
Product Code. 15530112
  • £23.83 / Pack of 10
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
  • Comes with flat bottom shape to allow use as weighing boat
  • Available in white color
  • Available in individual packs for one time use
  • Includes certificate of processing for sterilization
Specifications
Specifications
Spoon
Not Autoclavable
Flat
Single-use
White
Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders
Polystyrene
Certificate of processing for sterilization
5 mL
Sterile
FDA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Sterileware™ Economy Sample Spoons > Bowl capac.: 0.17 oz. (5mL)