Learn More
Nickel Electro™ Stainless Steel Flask Clip
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
Brand: Nickel Electro™ FC0050
This item is not returnable.
View return policy
Description
- Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
- No sharp edges to cut or score glassware
- Can be autoclaved
- Plastic base
- Easy to assemble and remove Flask Clips from Perforated tray
Specifications
50 mL Conical Flask | |
Flask Clip | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Flask Clip > For Use With: 50mL Conical Flask
Spot an opportunity for improvement?Share a Content Correction