Learn More
Contec™ ProChlor

Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.
Brand: Contec™ SBC102PC
204.22 GBP valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Ensures cleanroom is free from contamination and particulates.
A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.
Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
- Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
- Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
- Sterile version available for Grade A and B cleanrooms
- Contains no quaternary ammonium or surfactant so it's very low residue
- No hazard classification means only basic PPE required and no special disposal is required
- Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom

Specifications
Disinfectant | |
3 to 6 | |
Colorless | |
Grade A and B cleanrooms | |
Fluid | |
Limited-use | |
No | |
1 L |
Liquid | |
Sterile | |
Not Autoclavable | |
Clear | |
Capped Bottle | |
Filtered | |
6 x 1 L |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
241102945 | Certificate of Quality | 10/04/2025 | SBC102PC |
240602790 | Certificate of Quality | 22/11/2024 | SBC102PC |
230602480 | Certificate of Quality | 19/04/2024 | SBC102PC |
230502422 | Certificate of Quality | 01/02/2024 | SBC102PC |
230202326 | Certificate of Quality | 23/06/2023 | SBC102PC |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
18Hours
12Minutes
24Seconds
21
Your input is important to us. Please complete this form to provide feedback related to the content on this product.