missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate

SuperSignal™ West Atto Ultimate Sensitivity Substrate is an enhanced chemiluminescent (ECL) substrate for high attogram protein detection and provides the high level sensitivity and better signal-to-noise ratios compared to other commercially available horseradish peroxidase (HRP) substrates.

Brand:  Thermo Scientific™ A38556

523.98 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 16595740

  • £738.00 / 200mL
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sensitivity: low-femtogram to high-attogram
  • Stability: eight hours for working solution, one year for kit at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to four hours
  • Recommended primary antibody concentration: 1:1000 to 1:5000 dilution (0.2 to 1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:100000 to 1:250000 dilution (4 to 10 ng/mL)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
Nitrocellulose, PVDF
200 mL
Low-femtogram to high-attogram
SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide

• Substrate Solution, 100 mL
• Stable Peroxide Solution, 100 mL

Sufficient for 25 mini-gel blots; 2000 cm2 of membrane.

Store reagents at 2–8°C.
Chemiluminescence
Western Blot
SuperSignal™
Horseradish Peroxidase
25 Mini-Gel Blots, 2000 sq. cm of Membrane
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA403488 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberA38556
Lot NumberZL410827 Certificate TypeCertificate of Analysis Date24/12/2024 Catalog NumberA38556
Lot NumberZK400541 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA38556
Lot NumberZK409139 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA38556
Lot NumberZK409140 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA38556
Special Offers

Special Offers

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-31

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SuperSignal™ West Atto Ultimate Sensitivity Substrate > Quantity: 200 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.