Learn More
PURUS™ Synthetic Cleanroom Paper
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
Brand: PURUS™ LCIW 2034C
This item is not returnable.
View return policy
Description
- Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
- Propriatary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
- Unique precision cutting technology guarantees consistent sizing and cleanliness
- Produced in ISO 4 (Class 10) cleanroom
- Recyclable
- Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Synthetic | |
White | |
Autoclavable | |
Yes | |
250 Sheets/Pk., 20 Packs/Cs. | |
100 gsm |
No | |
210 x 148 mm | |
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments | |
Cleanroom Paper | |
A5 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
PURUS™ Synthetic Cleanroom Paper > White,A5,100gsm
Spot an opportunity for improvement?Share a Content Correction