Learn More
Thermo Scientific™ TheraPure™ Restriction Enzymes
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Brand: Thermo Scientific™ ER023T1
Description
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.
The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.
Features
- Convenient stand-alone enzymes for easy workflow optimization
- Animal-origin-free (AOF) raw materials used in manufacturing
- No risk of prion or animal virus contamination
- Reproducible and consistent results with validated quality testing
- Regulatory compliant—β-lactam-free manufacturing technology
- Suitable for translational research
- Facilitates transition to GMP during mRNA vaccine development process
- Scalable and available as OEM product
The exceptional quality of TheraPure products is supported by:
- Certificates of analysis
- Certificates of origin
- TSE/BSE statement

Specifications
20 U/μL | |
In vitro Transcription | |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Tango Buffer | |
37°C | |
3000 U, 150 μL | |
Restriction Enzyme | |
TheraPure™ | |
Liquid |
37°C | |
Store at -20°C. | |
Eam1104I | |
Yes | |
Yes | |
CTCTTC(1/4)^ | |
Drug Development | |
65°C |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.