Learn More
Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes
Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.
Brand: Wilmad-LabGlass™ 66000-0165
255.10 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Designed for routine use in most low to mid field NMR spectrometers
- One of the best OD tolerances in the industry
- Guaranteed to fit tightly in spinner turbine
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimension to ensure the success of your experiments
- WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Specifications
60 μm | |
ASTM E438 Type 1 Class B Glass | |
Bruker SampleJet Cap | |
600 MHz | |
0.43 mm | |
WG-1000-7-SJ |
178 mm | |
NMR Tube | |
5 mm | |
7 in. | |
100/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.