Learn More
Fisherbrand™ Triangular PTFE Stir Bars
Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container. The triangular PTFE stir bar provides stable stirring action for flat-bottom containers.
Brand: Fisherbrand™ 001.440
Description
- Stable, general-purpose magnetic stir bar
- Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container
- Autoclavable
Specifications
Autoclavable | |
White | |
40 x 14mm | |
14mm | |
PTFE | |
5/Pk. | |
Stir Bar |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
Containers & Vessels / Stirrer Hot Plates | |
14mm | |
40 mm | |
Triangle |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.