Learn More
Integra Biosciences™ Vacusip Aspiration System
Designed for convenient and affordable disposal of supernatant or excess liquids. Integra Biosciences™ Vacusip Aspiration System is a compact and ready-to-use vacuum aspiration system with an integrated silent vacuum pump for the aspiration of small volumes of liquids.
Brand:  Integra Biosciences™ 159010
Product Code. 11747312
  • £1114.30 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eliminates tedious handling and the risk of contaminating house vacuum system
  • The VACUSIP is an out of the box solution
  • No installation or additional equipment is required for immediate use
  • The integrated pump runs silently and stops automatically when the vacuum is established
  • Changing a filled collection bottle of VACUSIP is clean and convenient
  • VACUSIP is small, compact and fits on every bench.
  • Due to its independence from an external vacuum source or power supply, it can be relocated wherever needed
  • When working in a safety cabinet, only a minimum of tubing and cable is desirable
  • Due to the rechargeable battery in VACUSIP, a power supply cable is not needed
  • Hydrophobic filter protects the unit from damage and contamination by aerosols and liquids.
  • For easy decontamination, all parts that come in contact with liquids can be autoclaved
Applications include aspiration and removal of:
  • Small volumes of liquid out of microcentrifuge tubes or microtiter plates.
  • Supernatants after centrifugation steps (e.g. RNA/DNA extraction)
  • Washing solutions in Western blots and ELISA
  • Remaining quantities of reagents
  • Buffers of Western blot strips
  • Excess liquid from object slides and Petri dishes
  • Bacterial solutions
  • Cell staining solutions
Specifications
Specifications
Aspiration of small volumes of liquids
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Integra Biosciences™ Vacusip Aspiration System > 100-240VAC