missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Mini Pictograms Hazardous Labels
Brand: Brady™ 814019
14.83 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Vinyl | |
Permanent Acrylic | |
10 mm | |
-18°C to 60°C | |
DANGER for Aquatic Environment | |
Red |
MINI PIC1806-10X10-GHS09/96 | |
96/Pk. | |
Pipe markers and safety signs | |
148 mm | |
Diamond | |
First Aid Sign |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
27Hours
17Minutes
35Seconds
25
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ Vinyl Mini Pictograms Hazardous Labels > Danger for Aquatic Environment
Spot an opportunity for improvement?Share a Content Correction