missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WebSeal™ 96-Square Well Microplates with Precut Sealing Mat

Save time and minimize the risk of cross-contamination with convenient 96-Well Plate Kits packaged with pre-assembled glass or PTFE vials.

Brand:  Thermo Scientific™ 60180-K108

72.02 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 15102779

  • £81.86 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Save time and minimize the risk of cross-contamination with Thermo Scientific™ WebSeal™ 96-Well Plate Kits with Sealing Mats. Compatible with all 96-well-configured autosamplers, the ready-to-use kits consist of a comprehensive choice of 96-well microplates, pre-inserted glass or PTFE vials and silicone/PTFE sealing mats.

  • Convenient wellplate kits include pre-inserted glass or PTFE vials and 96-position sealing mats; some include cutting tool
  • Mats seal samples for storage
  • Plates are glass-inserted for greater inertness compared to pure PP plates
  • Unique cutting tool allows removal of individual samples with the seal in place, eliminating cross-contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Webseal
Microplate
96
Square
Constricted
45 mm
96 Square Well Plate with Sealing Mat - Precut
1 Pk.
Clear
Silicone, PTFE, Glass
1000 μL
1
1/Pk.
7.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ WebSeal™ 96-Square Well Microplates with Precut Sealing Mat > 45mm H; 1000uL; 7.6mm ID; Constricted profile; Pre-slit cap

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.