Learn More
Woodsafe™ Woodsafe Sharpsbin Holders
Accessories to Woodsafe sharpsbins and clinical waste bins. Holders and trolleys.
Brand: Woodsafe™ 5005028
160.55 GBP valid until 2025-05-30
Use promo code "24004" to get your promotional price.
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Steady holders and trolleys
Specifications
White Metal String | |
20.5 x 6.5 x 11 cm | |
Holder |
Woodsafe Sharpsbin Holders | |
0.5 L Woodsafe containers |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Lower climate impact and ACT label