Learn More
3,4-(Methylenedioxy)aniline, 98+%, Thermo Scientific Chemicals
Description
3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,4-(Methylenedioxy)aniline is an pharmaceutically important aniline derivative. It undergoes N-alkylation with cyclic secondary alkylamines in the presence of Shvo catalyst to yield N-arylpyrrolidines. 3,4-(Methylenedioxy)aniline was used in the synthesis of γ-glutamylanilides.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong acids, acid chlorides, acid anhydrides. Air & Light Sensitive. Stable under recommended storage conditions.

Specifications
Specifications
Melting Point | 40°C to 45°C |
Boiling Point | 144°C (16 mmHg) |
Flash Point | 142°C (287°F) |
Quantity | 10 g |
UN Number | UN2811 |
Beilstein | 4919 |
Sensitivity | Air and light sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 137.14 |
Percent Purity | ≥98% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10144139 | Certificate of Analysis | 30/08/2024 | A12975.18,A12975.09 |
10243794 | Certificate of Analysis | 21/10/2022 | A12975.18,A12975.09 |
10221637 | Certificate of Analysis | 22/11/2021 | A12975.18,A12975.09 |
5017R23R | Certificate of Analysis | 22/11/2021 | A12975.18,A12975.09 |
10235961 | Certificate of Analysis | 22/11/2021 | A12975.18,A12975.09 |
Safety and Handling

- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.