missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Non Binding Microplates
Features long-term surface stability without degradation or leaching.
Brand: Greiner Bio-One 655901
This item is not returnable.
View return policy
Description
- Ultra low non-specific biomolecular binding properties (proteins, DNA, RNA)
- Long-term surface stability without degradation or leaching
- Higher assay sensitivity with reduced background
- Significantly increase assay sensitivity

Specifications
Free of detectable DNase/RNase, human DNA and pyrogens | |
8 x 12 | |
Clear | |
96 | |
Non-sterile | |
40 | |
40/Cs. | |
Chimney |
Standard | |
Not Autoclavable | |
Polystyrene | |
25 to 340 μL | |
Flat Bottom (F-well) | |
10 | |
-20°C to +60°C | |
128 x 85 x 14 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One 96-well Non Binding Microplates > Flat bottom
Spot an opportunity for improvement?Share a Content Correction