missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aerosol-tight Rotor FA-24x2

Eppendorf™ Aerosol-tight Rotor FA-24x2 is suitable for applications in the field of molecular and cell biology as well as protein biochemistry.

Brand:  Eppendorf™ 5495500006

601.98 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


Product Code. 15831545

  • £691.60 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Rotor for Eppendorf Centrifuge 5425
  • Max. capacity: 24 x 1.5/2.0mL tubes
  • Aerosol-tight with Eppendorf QuickLock™ lid
  • Max. speed: 21,300 x g (15,060rpm)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum
24 x 1.5/2 mL
Aerosol-tight QuickLock rotor lid
Aerosol-tight
Fixed Angle Rotor
Eppendorf 5425
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Aerosol-tight Rotor FA-24x2 > Capacity: 24 x 1.5/2.0mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.