Learn More
IL-15 Monoclonal Antibody (34559), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523561
235.34 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2, recombinant mouse IL-15, or rhIL-21 is observed.
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Flow Cytometry.
Specifications
IL-15 | |
Monoclonal | |
PE | |
IL15 | |
AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15 | |
Mouse | |
Protein A/G | |
RUO | |
3600 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
34559 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P40933 | |
IL15 | |
E. coli-derived recombinant human IL-15 Asn49-Ser162 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.