Learn More
Applied Biosystems™ SYBR™ Green Universal Master Mix
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Brand: Applied Biosystems™ 4344463
59.95 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.
Description
Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.
- SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
- AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
- dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
- Proprietary buffer enhancements ensure performance and reliability
- Premixed components stored at 2° to 8°C significantly reduce assay setup time
Maximum Flexibility and Convenience:
- Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
- SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
- The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
- Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
- SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
- Store at -20°C
Order Info
Shipping Condition: Wet Ice

Specifications
SYBR™ Green PCR Master Mix in a 1 mL vial. Sufficient reagents provided for 40 reactions based on a 50 μl reaction volume. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Concentrated | |
High | |
AmpliTaq Gold DNA Polymerase | |
Fluorescence Intensity | |
SYBR Green | |
SYBR™ | |
1 x 1 mL | |
40 Reactions of 50 μL | |
Gene Expression | |
Built-In Hot Start | |
DNA |
SYBR | |
Tube | |
qPCR | |
Standard | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System, GeneAmp™ 5700 | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Wet Ice | |
Not Target-Specific | |
2X | |
40 Reactions | |
1 x 1 mL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2502137 | Certificate of Analysis | 15/03/2025 | |
2408134 | Certificate of Analysis | 31/10/2024 | 4344463_LITL18 PROMO,4344463 |
2410135 | Certificate of Analysis | 22/10/2024 | 4344463_LITL18 PROMO,4344463 |
2406133 | Certificate of Analysis | 18/06/2024 | 4344463_LITL18 PROMO,4344463 |
2403132 | Certificate of Analysis | 12/04/2024 | 4344463_LITL18 PROMO,4344463 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.