Learn More
Specac™ Atlas™ Hydraulic Press Automatic 15T
Brand:  Specac™ GS25430
Encompass_Preferred
Product Code. 15270348
  • £14267.11 / Each
Explore more special offers
Description
Description
This Atlas Power 15T Press is a power assisted hydraulic press that operates at up to 15 Tons. It has been designed to handle a wide variety of pressing applications including XRF and IR sample preparation. This press has a microprocessor controlled pressure application and release, Touch Screen operation and can accommodate samples up to 200mm in diameter. They can also be used with Specac Heated Platens GS15515 and Specac Evacuable Pellet Dies. Supplied with UK/EU Plug.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Specac™ Atlas™ Hydraulic Press Automatic 15T