Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
Brand:  Bel-Art™ H19558-0000
 View more versions of this product
Product Code. 15379658
  • £47.50 / Pack of 12
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
Autoclavable
4.7 to 6mm
Reducing connector
Barbed Fitting
Polypropylene
For 3/16 in. to 1/4 in. tubing (4.7 to 6mm)
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector > Barbed Fittings, Size: 3/16 x 1/4 in.