Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.)
Brand:  Bel-Art™ 06458-50
 View more versions of this product
Product Code. 12429620
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
Autoclavable
6 to 12.7mm
Reducing connector
Barbed Fitting
Polypropylene
For 1/4 in. to 1/2 in. tubing (6 to 12.7mm)
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector > Barbed Fittings, Size: 1/4 to 1/2 in.