Learn More
Bel-Art™ Clavies™ High-Temperature Autoclave Bags
Withstand high temperatures to reduce autoclave time.
£76.65 - £366.00
Specifications
Product Type Autoclave Bags
Autoclavable Autoclavable
Certifications/Compliance Dart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Color Clear
Disposable Single-use
View More Specs
Includes: Bag closures and holders sold separately.
Products 9
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
11774286
View Documents
Bel-Art™
H13182-1010
10 x 10 in.
£87.90
Pack of 100
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
13293529
View Documents
Bel-Art™
H13182-0815
15 x 8 in. (38.1 x 20cm)
£89.20
Pack of 100
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
15421690
View Documents Promotion Details
Bel-Art™
H13182-1012
12 x 10 in. (30.48 x 25.4cm)
£76.65
Pack of 100
Estimated Shipment: 30-06-2025
to see stock.
Add to basket
15520513
View Documents Promotion Details
Bel-Art™
H13182-1015
15 x 10 in. (38.1 x 25.4cm)
£77.65
Pack of 100
In Stock
to see stock.
Add to basket
10768895
View Documents Promotion Details
Bel-Art™
H13185-2436
36 x 24 in. (91 x 61 cm)
£366.00
Pack of 100
In Stock
to see stock.
Add to basket
10799465
View Documents
Bel-Art™
H13185-2430
30 x 24 in. (76 x 61 cm)
£321.00
Pack of 100
In Stock
to see stock.
Add to basket
10490955
View Documents Promotion Details
Bel-Art™
H13185-1224
24 x 12 in. (61 x 30 cm)
£127.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
13273529
View Documents Promotion Details
Bel-Art™
H13182-0812
12 x 8 in. (30 x 20 cm)
£81.60
Pack of 100
In Stock
to see stock.
Add to basket
12695457
View Documents Promotion Details
Bel-Art™
H13182-0810
10 x 8 in. (25.4 x 20cm)
£77.75
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Withstand high temperatures to reduce autoclave time
  • Suitable for discarding Petri dishes, membrane filters, multi-well culture plates, pipets and more
  • Rugged, transparent, 0.05 mm (2 mil) thick polypropylene
  • Steam autoclavable at 275°F (135°C)
Specifications
Specifications
Autoclave Bags
Dart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Single-use
2 mil
Autoclavable
Clear
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Bel-Art™ Clavies™ High-Temperature Autoclave Bags