missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BIX-01294, 95%
Brand: Thermo Scientific™ 467831000
177.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
BIX-01294 | |
2-(Hexahydro-4-methyl-1 H-1,4-diazepin-1-yl)-6,7-dimethoxy-N-[1-(phenylmethyl)-4-piperidinyl]-4-quinazolinamine | |
C28 H38 N6 O2 | |
Solid | |
95% |
935693-62-2 | |
100 mg | |
490.65 | |
Reagent |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
344429 | Certificate of Analysis | 16/10/2020 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ BIX-01294, 95% > 100 mg
Spot an opportunity for improvement?Share a Content Correction