missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bsm DNA Polymerase, large fragment (8 U/μL)

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities.

Brand:  Thermo Scientific™ EP0691

Product Code. 10364530

  • £85.00 / 1600 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities. Sourced from E. coli cells with a cloned polA gene from Bacillus smithii, Bsm DNA Polymerase, Large Fragment, features strong strand displacement activity and is active in a wide range of temperatures (30–63°C), with an optimum of activity at 60°C. With high functional similarity to Bst DNA Polymerase, Large Fragment, this enzyme can serve as alternative in most applications including loop-mediated isothermal amplification (LAMP).

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities. Sourced from E. coli cells with a cloned polA gene from Bacillus smithii, Bsm DNA Polymerase, Large Fragment, features strong strand displacement activity and is active in a wide range of temperatures (30–63°C), with an optimum of activity at 60°C. With high functional similarity to Bst DNA Polymerase, Large Fragment, this enzyme can serve as an alternative in most applications, including loop-mediated isothermal amplification (LAMP).

Applications
Bsm DNA Polymerase, Large Fragment, may be used in a wide variety of applications including:
• Loop-mediated isothermal amplification (LAMP)
• Whole genome amplification (WGA)
• Ramification amplification (RAM)
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stand-alone enzyme
Bsm DNA Polymerase

• Bsm DNA Polymerase, Large Fragment (200 μL at 8 U/μL)
• 10X Bsm Buffer (1.25 mL)

Store at –15 to –25°C.

Dry Ice
30 to 60 minutes
LAMP, RT-LAMP, WGA, RAM
1,600 units
60°C
8 U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2809178 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0691
Lot Number2826205 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0691
Lot Number2856733 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0691
Lot Number2857724 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0691
Lot Number2923741 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberEP0691
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Bsm DNA Polymerase, large fragment (8 U/μL) > 1600 U

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.