Learn More
Invitrogen™ NativePAGE™ Sample Buffer (4X)
A solution used to solubilize native protein samples prior to use with the NativePAGE Gel System
Brand: Invitrogen™ BN2003
45.62 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
The buffer is used along with 10% DDM or 5% Digitonin detergents to solubilize hydrophobic or membrane proteins. This buffer is also included in the NativePAGE Sample Prep Kit. The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice

Specifications
Store at 2°C to 8°C | |
4 X | |
PAGE Sample Buffer | |
NativePAGE™ | |
Wet Ice |
Sample Loading Buffers | |
Native-PAGE | |
NativePAGE™ Gels | |
10 mL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.