Learn More
Invitrogen™ NativePAGE™ Running Buffer (20X)
Used to make the NativePAGE Cathode and Anode Running Buffers
Marke: Invitrogen™ BN2001
161.17 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Beschreibung
For use with an XCell SureLock™ Mini Cell when running NativePAGE Gels.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Bestellinformation
Shipping Condition: Room Temperature

Spezifikation
Store at room temperature | |
20 X | |
PAGE Running Buffer | |
NativePAGE™ | |
Room Temperature |
Running Buffers | |
Native-PAGE | |
NativePAGE™ Gels | |
1 L |
Produktvorschläge
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only. Not for use in diagnostic procedures.