Learn More
Mettler Toledo™ Cyanide Half Cell Electrode
Brand:  Mettler Toledo™ 51107681
1075.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11977701
  • £1120.00 / Each
Estimated Shipment: 11-06-2025
Explore more special offers
Add to basket
Specifications
Solid State Membrane
0°C to 80°C
4 to 13
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Cyanide Half Cell Electrode > Membrane: Solid State