Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand: Thermo Scientific™ D8825
212.80 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
High Temp, Ultrapure, Oxygen Removal Cartridge | |
Ultrapure and Oxygen Removal Resin | |
Deionizer Cartridge |
Mixed Bed, Ultrapure, Oxygen and Organic Removal | |
B-Pure™ |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
- Multiple Manufacturers