Learn More
Merck Millicell HY 5-Layer Cell Culture Flask, T-1000, Sterile
The T-1000 sterile, high yield cell culture flasks provide a consistent, quality growth environment across all layers, with the same volume of media in each layer and the same media requirements as traditional T-flasks.
Brand: Merck PFHYS1008
Description
- Consistent, high cell yields and viability, uniform health across all layers
- Same media requirements (volume per unit area) as traditional T-flasks
- Uniform culture conditions without media leaking or spillover between layers
- Save on time spent passaging cells
- Save on cost of media, cells and reagents
- An easy transition from single-layer cell culture flasks
- Easy media removal from flask with ergonomic cap location
- Fits under microscopes for monitoring cell health and confluency
- Stackable design saves incubator space
- Bar-coding available
- Frosted areas on flask allow for easy marking and numbering

Specifications
200 mL to 300 mL (recommended media volume) | |
Polystyrene | |
2 in. | |
8.7 in. | |
8 Pk. | |
Cell Culture Flask | |
4.7 in. | |
Sterile | |
5-layers |
Red | |
Cell growth | |
51 mm | |
222 mm | |
1/Bag | |
45°C (max.) | |
120 mm | |
1000 cm2 |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.