missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Gibco™ Distilled Water is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, Gibco™ Distilled Water features:
Cell culture quality
Proven performance
Use in cell culture and molecular biology applications
Cell culture quality
Gibco™ Distilled Water is prepared by distillation and filtration (0.1μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.
Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
Use in cell culture and molecular biology applications
Gibco™ Distilled Water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Distilled Water is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Distilled Water product made in our Grand Island facility (15230-204). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Cell culture quality
Gibco™ Distilled Water is prepared by distillation and filtration (0.1μm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin and bioburden are tested daily; metals are tested monthly.
Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.
Use in cell culture and molecular biology applications
Gibco™ Distilled Water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions and diluting buffers.
Product Use
For Research Use Only. Not for use in diagnostic procedures.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Distilled Water is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Distilled Water product made in our Grand Island facility (15230-204). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Greener Choice Claims
- Environmental benefits include:
- Renewable Materials
- Recycled Content

Specifications
Specifications
Description | Distilled Water, 10 x 500mL, Bottle(s), Membrane-Filtered Purification, Sterile, 15 to 30°C Storage Conditions, Ambient, Fewer Resources Used/Less Waste, ISO 13485/cGMP/FDA Certified, This Water is Also Bottled as Gibco™ Distilled Water |
Sterility | Sterile |
Treatment(s) | Not DEPC-treated |
Certificate of Analysis | ISO 13485, cGMP, FDA |
Quantity | 10 x 500 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Distilled Water >
Spot an opportunity for improvement?Share a Content Correction