Learn More
Fisherbrand™ Reusable Amber Glass Media Bottles
Shop All Fisherbrand ProductsDescription
Fisherbrand™ Reusable Amber Glass Media Bottles with cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-free, replaceable polypropylene pour ring.
- Permanent white enamel marking spot
- GL 45 screw thread
- Linerless
- Autoclavable (to 140°C)
- Amber borosilicate glass
- Polypropylene cap

Specifications
Specifications
Capacity (Metric) | 250 mL |
Color | Amber |
For Use With (Application) | Media and reagent storage, Holding and dispensing fluids, Chemical storage |
Includes | Screw Cap, Pouring Ring |
Material | Borosilicate Glass (3.3) |
No. per Case | 10/Cs. |
Screw Color | Blue |
Screw Material | Polypropylene |
Sterility | Non-sterile |
Closure Color | Blue |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
35Hours
21Minutes
21Seconds
57New Offer Save Up to 70% -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.