Learn More
MP Biomedicals™ GeneClean Kit
GeneClean™ Kit, Solution or Gels
Brand:  MP Biomedicals™ 111001200
Product Code. 11363099
  • £295.00 / 200 preps
Explore more special offers
Description
Description
  • The GENECLEAN™ Kit includes Glassmilk™ slurry that allows flexibility of reaction size and volume.
  • The GENECLEAN™ kit simplifies the process of purifying DNA into three easy steps: Bind,Wash and Elute.
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
  • Isolate DNA from TAE buffered agarose gels and solutions
  • The GENECLEAN™ Kit is used to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
New Wash/Ethanol Concentration must be ∽ 50%
Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away fromgenomic DNA.
Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Solution or Gels
Store at Room Temperature (15-30°C). NaI solution must be kept away from light and open air sources to prevent oxidation. NaI is packaged under nitrogengas to further reduce chance of oxidation. Shelf life may be extended by storing at 4°C.
200 Preps
Kits and Assays
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ GeneClean Kit > 200 preps