Learn More
Gibco™ TrypLE™ Express Enzyme (1X), phenol red

Animal origin-free, recombinant enzyme
Brand: Gibco™ 12605028
60.64 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.
Description
Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE™ Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.
- Gentle On Cells
- urity increases specificity due to the action of single enzyme, reducing damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts
- Room-Temperature Stable for 24 months
- Ready to use
- Can be directly substituted for trypsin in existing protocols
- Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured in facilities compliant with cGMP manufacturing requirements, certified to the ISO 13485 standard, and registered with the FDA as medical device manufacturers.

Specifications
EDTA | |
Mammalian Cells | |
1 X | |
Liquid | |
Sterile-filtered | |
Energy efficient, Sustainable packaging | |
Gibco™ | |
Room Temperature Stable | |
Animal origin-free |
Storage conditions: 15°C to 30°C, protect from light Alternate storage conditions: 2°C to 8°C or -5°C to -20°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Animal Origin-free | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
Phenol Red, EDTA | |
Check and Record | |
500 mL | |
Room Temperature | |
In vitro Bioassay |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2658503 | Certificate of Analysis | 29/01/2025 | 12605010,12605028 |
2537004 | Certificate of Analysis | 29/01/2025 | 12605010,12605028 |
2742877 | Certificate of Analysis | 29/01/2025 | 12605010,12605028 |
2766542 | Certificate of Analysis | 29/01/2025 | 12605010,12605028 |
2744469 | Certificate of Analysis | 29/01/2025 | 12605010,12605028 |
Special Offers
-
Thermo Scientific Nunc
,
Thermo Scientific Nalgene
,
Gibco
,
Invitrogen
,
Thermo Scientific
Cell Biology Promotion
Discover how you can maximise your savings and boost your research to the next level with our top-of-the-line Gibco and Thermo Scientific products.Promo Code: 24071Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.