missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Modified Eagles Medium

Designed to support cell culture growth and biomanufacturing processes.

Brand:  Cytiva SH30081.FS

95.85 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.


 View more versions of this product

Product Code. 11988075

  • £113.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
0.1 μm Sterile-filtered
No Glutamine, No Sodium Pyruvate
ISO 9001:2000
DMEM/High glucose without L-glutamine, sodium pyruvate
Glucose
6 x 500 mL
2° to 8°C, away from light
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ Dulbecco's Modified Eagles Medium > DMEM/High; Without L Glutamine and Sodium Pyruvate; 6 x 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.