Learn More
Ibrutinib-biotin, MedChemExpress
MedChemExpress Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.
Brand: Medchem Express HY-100342
Description
Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.Specifications
Ibrutinib-biotin | |
Colorless | |
C56H80N12O9S | |
DMSO : 50 mg/mL (45.56 mM; Need ultrasonic) | |
1097.37 | |
98.04% | |
Oil |
1599432-18-4 | |
Research | |
10 mg | |
O=C(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@@H]1SC[C@H](N2)[C@@H]1NC2=O)=O)=O)NCCCCN(C)C/C=C/C(N3CCC[C@@H](N4N=C(C5=CC=C(OC6=CC=CC=C6)C=C5)C7=C(N)N=CN=C74)C3)=O | |
1097.37 | |
Research | |
Cancer-Kinase/protease |
Product Suggestions
Customers who viewed this item also viewed.