Learn More
Emma Safety Footwear Metric Safety Shoes
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
Brand: Emma Safety Footwear MM50654036
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist Outsole
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
White | |
Hydro-Tec™ Sanitized Silver | |
36 |
Elastic | |
Metric | |
Microfiber | |
Steel |
Product Suggestions
Customers who viewed this item also viewed