missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TeenPrep™ Lysing Matrix D Tubes

TeenPrep™ Lysing Matrix D Tubes contain 1.4 mm zirconium-silicate spheres and are used to grind soft animal and plant tissues, cultured cells and insects/worms.

£27.35 - £4450.00

Specifications

Bead Type 1.4 mm Zirconium-silicate Spheres
Sample Type Animal Tissues, Plants, Cultured Cells, Insects, Worms
DNase Dnase-free
For Use With (Application) Nucleic Acid Isolation and Purification, Homogenization
Matrix Lysing Matrix D
View More Specs

Products 16
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12731461
View Documents
MP Biomedicals™
116933050
50 x 15 mL
£746.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
15709200
View Documents
MP Biomedicals™
116933005
5 x 15 mL
£81.20
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
11461555
View Documents
MP Biomedicals™
116933025
25 x 15 mL
£248.00
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
15792462
View Documents
MP Biomedicals™
116953002
2 x 50 mL
£27.35
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
11451575
View Documents
MP Biomedicals™
116973025
25 x 4.5 mL Tubes
£198.00
Pack of 25
Please to purchase this item. Need a web account? Register with us today!
12323873
View Documents
MP Biomedicals™
116983001
1 x 96-Well Plate
£188.00
Each
Please to purchase this item. Need a web account? Register with us today!
11412420
View Documents
MP Biomedicals™
116913050
50 x 2 mL Tubes
£320.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
11400506
View Documents
MP Biomedicals™
116953010
10 x 50 mL
£134.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11461575
View Documents
MP Biomedicals™
116973050
50 x 4.5 mL Tubes
£277.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
15232870
View Documents
MP Biomedicals™
116983010
10 x 96-Well Plate
£1825.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11422420
View Documents
MP Biomedicals™
116913100
100 x 2 mL Tubes
£516.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
12761461
View Documents
MP Biomedicals™
116953050
50 x 50 mL
£557.00
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
11471575
View Documents
MP Biomedicals™
116973100
100 x 4.5 mL Tubes
£592.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
11432420
View Documents
MP Biomedicals™
116913500
500 x 2 mL Tubes
£1815.00
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
17466483
View Documents
MP Biomedicals™
116953100
100 x 50 mL
£968.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
17476483
View Documents
MP Biomedicals™
116953500
500 x 50 mL
£4450.00
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TeenPrep™ Lysing Matrix D, tubes can be used with FastPrep instruments or similar bead beater. The grinding matrix is chemically inert and will not bind nucleic acids.

  • Tube size: 15 mL
  • Sample Types: Soft tissues such as brain, liver, kidney, lung and spleen; plant tissues such as leaves, roots and fruits; cultured cells; whole insects; nematodes
  • Matrix Composition: 1.4 mm zirconium-silicate spheres
Specifications

Specifications

1.4 mm Zirconium-silicate Spheres
Dnase-free
Lysing Matrix D
Animal Tissues, Plants, Cultured Cells, Insects, Worms
Nucleic Acid Isolation and Purification, Homogenization
Rnase-free
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ TeenPrep™ Lysing Matrix D Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.