missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MycoSEQ™ Mycoplasma Detection Kits

The MycoSEQ Mycoplasma Detection Kit is a real-time mycoplasma PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7 for use as a nucleic acid amplification technique (NAT).

Brand:  Applied Biosystems™ 4460626

 View more versions of this product

Product Code. 30000672

  • £4190.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

MycoSEQ Mycoplasma Detection Kit
• Power SYBR Green PCR Master Mix (2X), store at -15°C to -25°C on receipt, 2°C to 8°C after first use, protected from light
• Mycoplasma Real-Time PCR Primer Mix (10X), -15°C to -25°C on receipt, 2°C to 8°C after first use
• Discriminatory Positive/Extraction Control, -15°C to -25°C
• Negative control, -15°C to -25°C on receipt, 2°C to 8°C after first use

PrepSEQ 1-2-3 Mycoplasma Nucleic Acid Sample Extraction Kit
• Cell Fractionation Buffer, store at 2°C to 8°C
• RNase Cocktail Enzyme Mix, -15°C to -25°C
• Lysis Buffer, 18°C to 25°C
• Wash Buffer Concentrate, 18°C to 25°C
• Elution Buffer, 18°C to 25°C
• Proteinase K Buffer, 18°C to 25°C
• Magnetic Particles, 18°C to 25°C
• Proteinase K, -15°C to -25°C

QS5 0.1 mL Block or 7500 FAST
MycoSEQ™
100 Reactions
Detection for BioProduction process media, cells, raw materials
SYBR Green
Assay Kit and Sample Preparation
Mycoplasma Detection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ MycoSEQ™ Mycoplasma Detection Kits > Assay Kit and Sample Preparation

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.