missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitrocellulose stripping buffer (10X)

Brand:  Thermo Scientific Chemicals J62541.AP

71.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15424909

  • £71.76 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for removal of secondary antibody after detection of highly abundant targets. Nitrocellulose Stripping Buffer may perform well with nitrocellulose membranes or antibody-dye conjugates. It is also employed for nucleic acid and protein related blotting works.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot Stripping Buffer
Liquid
500 mL
10X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX20K558 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberJ62541.AP,J62541.AK
Lot NumberU01K545 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog NumberJ62541.AP,J62541.AK
Lot NumberP17K529 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ62541.AP,J62541.AK
Lot NumberM16K539 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberJ62541.AP,J62541.AK
Lot NumberM16K540 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberJ62541.AP,J62541.AK
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Nitrocellulose stripping buffer (10X) > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only