Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Brand: Thermo Scientific™ D1-MTC
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Micro well; 15 (1X) teeth; 1.0mm thick | |
Owl | |
15 |
Owl™ D2 Wide Gel Electrophoresis System | |
Comb | |
1 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.