Learn More
Fisherbrand™ Glass Screw Neck Specimen Vials
Store specimens securely with squat or tall form glass vials. Fisherbrand™ Glass Screw Neck Specimen Vials are available in a variety of sizes. Fisherbrand™ polypropylene screw caps are sold separately.
£59.45 - £276.00
Specifications
Color Clear
Material Neutral Glass (Type 1B)
Closure Type Screw Cap
View More Specs
Products 9
Product Code Brand Description Volume (Metric) Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Price Quantity & Availability  
15364779
View accessory products View Documents Promotion Details
Fisherbrand™
Squat form 28 mL
£61.55
Pack of 140
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15394779
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 7 mL
£107.00
Pack of 399
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15304789
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 10.5 mL
£168.00
Pack of 399
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15354779
View accessory products View Documents Promotion Details
Fisherbrand™
Squat form 14 mL
£59.45
Pack of 190
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15314789
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 14 mL
£131.06
Pack of 264
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15324789
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 21 mL
£120.74
Pack of 231
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15344779
View accessory products View Documents Promotion Details
Fisherbrand™
Squat form 7 mL
£67.05
Pack of 264
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15374779
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 1.75 mL
£276.00
Pack of 882
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15384779
View accessory products View Documents Promotion Details
Fisherbrand™
Tall form 3.5 mL
£205.00
Pack of 666
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
  • Manufactured from clear Type 1B neutral glass
  • Squat and tall form options with standard neck sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials sold separately
Specifications
Specifications
Clear
Neutral Glass (Type 1B)
Screw Cap