Learn More
PAR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533527
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%) Marmoset, Mouse, Rat, Hamster (94%) Gorilla, Horse, Pig (89%).
Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.
Specifications
PAR2 | |
Polyclonal | |
Unconjugated | |
F2rl1 | |
coagulation factor II (thrombin) receptor-like 1; coagulation factor II receptor-like 1; F2R like trypsin receptor 1; F2RL1; Gpcr11; Gpr11; G-protein coupled receptor 11; PAR2; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; proteinase-activated receptor-2; Proteinase-activated receptor-2 G protein-coupled receptor 11; Proteinase-activated receptor-2, G protein-coupled receptor 11; thrombin receptor-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P55085 | |
F2rl1 | |
Synthetic 18 amino acid peptide from 2nd extracellular domain of human F2RL1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.