Learn More
Buerkle™ Polyethylene Bung
Manufactured with polyethylene for strength, durability and flexibility. Buerkle™ Polyethylene Bungs are available in a wide range of sizes.
Brand:  Buerkle™ 9760-1926
 View more versions of this product
Product Code. 11888172
  • £1.05 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Hollow stopper allows for use of tubes or funnels
  • Sealing rings provide a tight seal
  • Suitable for foodstuff
  • Octagonal shape facilitates easy opening
  • Polyethylene provides good chemical resistance
Specifications
Specifications
For Foodstuffs
White, Red
19 mm
Bung
19/26
16.4 mm
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Polyethylene Bung > Standard Taper Joint:19/26