Learn More
Masterflex™ Polypropylene Luer Fitting
Featuring a variety of fitting options, these Luer fittings are ideal for many applications including syringes or small tubing. Cole-Parmer™ Polypropylene Luer Fitting are available with standard, wide, and low-profile barbs.
Brand:  Masterflex™ 45508-56
 View more versions of this product
Product Code. 15116116
  • £125.00 / Pack of 25
Estimated Shipment: 07-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Three barb styles: standard, wide-bore, and low-profile.
    • Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
    • Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
  • Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
  • All parts meet tight dimensional tolerances for increased reliability and process purity.
  • Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
  • Pack of 25
Specifications
Specifications
ISO 594-1 standards
Female x Female x Female
Luer Fitting
Polypropylene
25 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Polypropylene Luer Fitting > For female x female x female