missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Flat Bottom Plate, 96-well

This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.

Brand:  Invitrogen™ EPX-88182-000

18.74 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 17806667

  • £20.32 / Pack of 96

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.

This plate is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.

Note that the reusable lid (Cat. No. EPX-88104-000) is sold separately.

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

Related products

  • ProcartaPlex Microplate Lid (Cat. No. EPX-88104-000)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Without Lid
Black
Plate
96
Hydrophobic
96 tests
ProcartaPlex
Single-use
Polystyrene
Non-sterile
Flat Bottom (F-well)
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number445527-000 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number
Lot Number439608-000 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number
Lot Number438647-000 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog Number
Lot Number433165-000 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberEPX-88182-000
Lot Number432501-000 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberEPX-88182-000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ProcartaPlex™ Flat Bottom Plate, 96-well > 1 Plate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.